Learn More
Restek™ Capillary Installation Gauge for Agilent GCs
SDP
Catalog No. 06729309 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With:
Varian GCs, 0.0625 in. Ferrules
Catalog No. For Use With
06-729-309 Varian GCs, 0.0625 in. Ferrules
1 options
Catalog No. 06-729-309 Supplier Restek™ Supplier No. 22335
Add to Cart
Edge
Add to Cart
Seats graphite ferrules onto column for consistent installations.
  • Made from high-quality stainless steel
  • Prevents crushed column ends
  • For 1/16 in. ferrules and fittings
Part No. 21399
  • For Agilent GCs
Part No. 22335
  • For Bruker/Varian GCs
Specifications
Type Gauge
For Use With Varian GCs, 0.0625 in. Ferrules