Learn More
Restek™ Capillary Nuts for Agilent™ 5890/6890/7890 GCs
SDP
Catalog No. p-3181058 Shop All Restek Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Compact Agilent-style Ferrules
Standard 1/16 in. Ferrules
Material:
Brass
Stainless Steel
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Material
06-711-784 Compact Agilent-style Ferrules Brass
06-718-135 Standard 1/16 in. Ferrules Stainless Steel
06-718-727 Standard 1/16 in. Ferrules Brass
06-718-730 Compact Agilent-style Ferrules Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-711-784 Supplier Restek™ Supplier No. 21878
Only null left
Add to Cart
Add to Cart
Available in brass or stainless steel.
  • Capillary column nuts
  • For use with Agilent GCs
Specifications
Type Capillary Nut
Material Brass
For Use With (Equipment) Compact Agilent-style Ferrules