Learn More
Restek™ CarboFrit™ Puller/Inserter Tool
SDP
Catalog No. 06711699 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Packaging Quantity:
1 Ea.
Catalog No. Packaging Quantity
06-711-699 1 Ea.
1 options
Catalog No. 06-711-699 Supplier Restek™ Supplier No. 21642
Shaped to insert and remove CarboFrit™ inserts
  • Bent end (90°) for inserting CarboFrit™ inserts
  • Hook end for removing CarboFrit™ inserts
Specifications
Type Inserter/Puller Tool
Packaging Quantity 1 Ea.