Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Carriage Motor Belt for Agilent 7673A/7673B
SDP

Catalog No. 06712034 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Packaging Quantity:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
06-712-034 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-712-034 Supplier Restek™ Supplier No. 22692
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Agilent 7673A, 7673B, and 7673C Autosamplers.

  • Meets original manufacturer's performance

Specifications

Type Carriage Motor Belt
Packaging Quantity 1 Each
For Use With Agilent 7673 A/7673 B/7673 C Autosamplers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.