Learn More
Restek™ Ceramic Scoring Wafers
SDP
Catalog No. 06717592 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Packaging Quantity:
5/Pk.
Catalog No. Packaging Quantity
06-717-592 5/Pk.
1 options
Catalog No. 06-717-592 Supplier Restek™ Supplier No. 20116
Varied edges for multiple tasks
  • Four straight scoring edges for cutting fused silica tubing
  • Four serrated edges for cutting MXT™ metal capillary columns
Specifications
Type Scoring Wafer
Packaging Quantity 5/Pk.