Description
- Chromosorb P support can hold up to 30 weight% of liquid stationary phase, making it the highest loading support available
- Chromosorb G support is the hardest support available and has the lowest surface area of all the diatomaceous earth supports
- Chromosorb W support is a flux-calcinated diatomite. This solid support is very fragile but offers the highest inertness of all diatomaceous earth supports.
Specifications
Specifications
| Particle Size | 100/120 Mesh |
| Quantity | 100 g |
| Type | Packing Material |
| Material | Chromosorb PNAW |