Learn More
Restek™ Column End-Fittings
SDP
Catalog No. p-3217784 Shop All Restek Corporation Products
Change view
Click to view available options
For Use With (Equipment):
4.6 mm Column, Valco and Parker Fittings
Valco and Parker Fittings
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
06-712-649 Valco and Parker Fittings
06-712-648 4.6 mm Column, Valco and Parker Fittings
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-712-649 Supplier Restek™ Supplier No. 25078
Only null left
Add to Cart
Add to Cart
These 0.25 in. compression end-fittings are compatible with 0.0625 in. connecting tubing using Valco and Parker fittings.
  • Fittings with distribution cone are intended for use on 3.0 to 4.6mm ID HPLC columns
  • Fittings with flat bottom are intended for use on 1.0 to 3.0mm ID HPLC columns
Specifications
Type Column End-Fitting
For Use With (Equipment) Valco and Parker Fittings