Learn More
Restek™ Deactivated, Universal Press-Tight Connectors
SDP
Catalog No. 06717764 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Packaging Quantity:
5
25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
06-717-764 5
06-717-765 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-717-764 Supplier Restek™ Supplier No. 20429
Add to Cart
Add to Cart
Universal fit for use with 0.1 to 0.53mm ID Restek columns (0.33 to 0.74mm OD).
  • Deactivated fused quartz helps eliminate activity for complete inertness along the GC flow path
  • Connect a guard column to an analytical column
  • Repair a broken column
  • Connect a column outlet to a transfer line
Specifications
Type Connector
Packaging Quantity 5
For Use With Fused Silica Capillary Column