Learn More
Restek™ Deactivated Wool
SDP
Catalog No. 06719304 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
Catalog No. Quantity
06-719-304 10 g
1 options
Catalog No. 06-719-304 Supplier Restek™ Supplier No. 24324
More inert than traditional wool
  • Use to vaporize a sample in a liner prior to introduction into a capillary column
Specifications
Packing Material None
Quantity 10 g