Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Detector Lamps
SDP

Catalog No. 06712918 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
Deuterium Lamp
Fluorescence Detector Flash Lamp
Lamp
For Use With (Equipment):
2487 Water Detector
486 Waters Detector
996, 2996 Water Detector
Agilent 8453 HPLC System
Agilent DAD G1315 A, G1365 A HPLC System
Agilent G1321 HPLC System
Agilent VWD G1314 A HPLC System
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
06-712-918 Deuterium Lamp 996, 2996 Water Detector
06-712-808 Lamp Agilent DAD G1315 A, G1365 A HPLC System
06-712-809 Lamp Agilent VWD G1314 A HPLC System
06-712-810 Deuterium Lamp Agilent 8453 HPLC System
06-712-919 Deuterium Lamp 2487 Water Detector
06-712-920 Deuterium Lamp 486 Waters Detector
06-712-811 Fluorescence Detector Flash Lamp Agilent G1321 HPLC System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 06-712-918 Supplier Restek™ Supplier No. 25408
Add to Cart
Add to Cart

Used for waters detectors or for Agilent HPLC Systems

  • Standard lamps have nominal 1,000-hour life

Specifications

Type Deuterium Lamp
For Use With (Equipment) 996, 2996 Water Detector
Includes 1 Chromatography Detector Lamp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.