Learn More
Restek™ Injector/Detector Plug Nut
SDP
Catalog No. 06718729 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Packaging Quantity:
2/Pk.
Catalog No. Packaging Quantity
06-718-729 2/Pk.
1 options
Catalog No. 06-718-729 Supplier Restek™ Supplier No. 21883
Cap off or check injector or detector for leaks, cleaning, flow-rate.
  • Use to cap off an injector to isolate leaks
  • Use to cap off a detector for thermal cleaning
  • Use to check a detector or make-up gas flow rate
  • Use to cap off a detector and prevent hydrogen from accidentally diffusing into the oven from an unused detector base
Specifications
Type Nut
For Use With (Application) Cap Off or Check Injector or Detector for Leaks, Cleaning, Flow-Rate.
Packaging Quantity 2/Pk.
For Use With HP GC