Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Direct Injection (DI) Liner Adapter for Agilent 5890 Packed Column GC
SDP

Catalog No. 06711510 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-711-510 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-711-510 Supplier Restek™ Supplier No. 21303
May include imposed supplier surcharges.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

0.25 in. graphite ferrule and stainless steel nut

For Agilent 5890 Packed Column GC.

  • Uses standard 0.0625 in. capillary nut and ferrules
  • Convenient wrench pad at base
  • Use with Agilent or Restek DI glass inserts or Restek DI Uniliner liners

Specifications

Product Type DI Liner Adaptor
Includes 0.25 in. graphite ferrule and stainless steel nμt
Quantity Each
For Use With (Equipment) For Agilent 5890 Packed Column GC
Liner Material Glass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.