Learn More
Restek™ Double Gooseneck Splitless Liners for Thermo™ TRACE™ and Focus™ SSL
SDP
Catalog No. 06711278 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Liner Material:
Glass (Closure Liner)
Quantity:
1/Ea.
Catalog No. Liner Material Quantity
06-711-278 Glass (Closure Liner) 1/Ea.
1 options
Catalog No. 06-711-278 Supplier Restek™ Supplier No. 20952
Double gooseneck design for increased performance with larger sample introductions.
4.0 mm ID Double Taper Inlet Liner for Thermo TRACE, 8000 Series and Focus GCs equipped with SSL inlets
Specifications
Quantity 1/Ea.
Length (Metric) 105 mm
Diameter (Metric) Outer 8 mm
Liner Material Glass (Closure Liner)