Learn More
Restek™ Easy Grip™ Manual Crimper
SDP
Catalog No. 06711290 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
20mm
Catalog No.
06-711-290 20mm
1 options
Catalog No. 06-711-290 Supplier Restek™ Supplier No. 23398
Feature tough, light, glass-filled plastic bodies with curved handles for outstanding hand comfort during use
  • Bottom-pull handle allows for a steadier hold than top-push handles
  • Easily viewed adjustment knob is located at the head of the product
  • Clearly shown + and - symbols, along with directional arrows, simplify adjusting the intensity of the crimp/decap needed
  • Knob is also designed to indicate when the crimp setting has been reached, when it fully contacts the body of the product
Specifications
Type Crimper