Learn More
Restek™ ECD/FID Dual-Purpose Make-Up Gas Fitting
SDP
Catalog No. 06711509 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Packaging Quantity:
1 Kit
Catalog No. Packaging Quantity
06-711-509 1 Kit
1 options
Catalog No. 06-711-509 Supplier Restek™ Supplier No. 21301
For use with Agilent 5890 GCs.
  • ECD/FID replacement fitting kit with flow manifold connection
  • Includes replacement fitting, 0.25 in. nut, Vespel/graphite ferrule, 0.062 in. nut, 0.4mm ID graphite ferrule, Siltek-treated guide
  • Quantity: 1 Kit
Specifications
Type Fitting
Packaging Quantity 1 Kit
Includes Replacement fitting, 0.25 in. nut, Vespel/graphite ferrule, 0.062 in. nut, 0.4 mm ID graphite ferrule, Siltek-treated guide
For Use With Agilent 5890 GCs