Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ FID Gauge Pack for Agilent 5890 GCs
SDP

Catalog No. 06802657 Shop All Restek Corporation Products
Encompass
Change view
Click to view available options
Packaging Quantity:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
06-802-657 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-802-657 Supplier Restek™ Supplier No. 22071
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Agilent 5890 GCs.

  • Pressure regulators and gauges for air and hydrogen
  • 1/8 in. bulkhead allows easy hookup to instrument
  • Rated for inlet pressures to 250psi (1,724 kPa)
  • Rated for outlet pressures of 0 to 60psi (0 to 414 kPa)

Specifications

Type FID Gauge Pack
Packaging Quantity 1 Each
For Use With Agilent 5890 GCs
Pressure Inlet: 250 psi (1724 kPa), Outlet: 0 to 60 psi (0 to 414 kPa)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.