Learn More
Restek™ FID/Injector Cleaning Kit
SDP
Catalog No. 06710502 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With:
Any Injection Port
Catalog No. For Use With
06-710-502 Any Injection Port
1 options
Catalog No. 06-710-502 Supplier Restek™ Supplier No. 20120
Includes
  • Nylon tube brushes (1/8 in., 3/16 in., 1/4 in.)
  • Pipe cleaner
  • Stainless steel tube brushes (3/8 in., 3/16 in., 1/4 in.)
  • Stainless steel surface brush
  • Stainless steel jet reamers
  • Emery cloth
Use with any injection port
  • Cleaning tool kit
Specifications
Type Tool Kit
For Use With Any Injection Port