Learn More
Restek™ FID/NPD Adapter Fitting
SDP
Catalog No. 06718136 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
06-718-136 Stainless Steel
1 options
Catalog No. 06-718-136 Supplier Restek™ Supplier No. 20884
Easy-to-use, sturdy, compact stainless steel fitting.
  • 0.0625 in. nut uses standard graphite or Vespel/graphite ferrules
  • Stable wrench pad
  • Includes 0.25- and 0.0625 in. stainless steel nuts, and 0.25 in. Vespel and 0.4mm ID graphite ferrules
Specifications
Type Adapter Fitting
Material Stainless Steel
For Use With (Equipment) Agilent GCs