Learn More
Restek™ Fingertight Nuts
SDP
Catalog No. 06711515 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Compact Agilent-Style Ferrules for HP GCs
Standard Ferrules for HP GCs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
06-711-515 Standard Ferrules for HP GCs
06-711-514 Compact Agilent-Style Ferrules for HP GCs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-711-515 Supplier Restek™ Supplier No. 21312
Rapidly tighten without wrenches and avoid stripped threads
  • Can be used with 0.25, 0.32, or 0.53mm ID columns
Specifications
Type Nut
For Use With (Equipment) Standard Ferrules for HP GCs