Learn More
Restek™ Gas-Tight SGE Syringes with PTFE Tip
SDP
Catalog No. 06712429 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
100 μL
250 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
06-712-429 250 μL
06-712-425 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-712-429 Supplier Restek™ Supplier No. 24741
Suitable for gases or liquids, for maximum inertness.
  • High accuracy of dispensed volumes
  • Interchangeable barrels, plungers, and tips extend performance and increase cost-effectiveness
Specifications
For Use With (Application) For gases or liquids, for maximum inertness
Product Type SGE PTFE Tip, Gas-Tight Syringes
Model 250F-LL-GT
Volume (Metric) 250 μL
Connection Type Luer Lock