Learn More
Restek™ GC Installation Kit
SDP
Catalog No. 06711520 Shop All Restek Corporation Products
Encompass_Preferred
Catalog No. 06-711-520 Supplier Restek™ Supplier No. 21325
Includes
  • Tubing cutter
  • One 1/8 in. x 1/4 in. reamer
  • One 1/4 in. x 1/8 in.brass tube end reducer
  • One 7/16 in. wrench
  • One 1/2 in. wrench
  • Four 1/8 in. brass tees
  • Ten 1/8 in. brass nuts
  • Ten brass front and back ferrules
  • 50 ft. (15.2m) of instrument-grade cleaned 1/8 in. copper tubing
All-in-one kit for ease of use
Contains tubing, fittings required to add an additional GC to a lab bench
Specifications
Type GC Installation Kit
Packaging Quantity 1 Kit