Learn More
Description
- High-Capacity Split Vent Trap reduces the release of hazardous materials from the capillary split vent into the lab
- ECD Vent Trap reduces the release of hazardous materials from the ECD vent into the lab
- Fittings: 1/8 in.
- Dimensions: 15.2 x 2.5cm (6 x 1 in.)
Specifications
Specifications
| Connection Type | 0.12 in. Fitting |
| Type | ECD Vent Trap |
| Includes | Connecting lines and mounting kit |
| Packaging Quantity | 1 Each |
| Length (English) | 6 in. |
| Length (Metric) | 15.2 cm |
| Diameter (English) Outer | 1 in. |
| Diameter (Metric) Outer | 2.5 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
