Learn More
Restek™ HPLC Piston Seal Insertion Tool
SDP
Catalog No. 06711542 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
06-711-542 1 Ea.
1 options
Catalog No. 06-711-542 Supplier Restek™ Supplier No. 21356
Add to Cart
Edge
Add to Cart
Use one end to remove your old seal, then simply slip your new seal on the other end and push it flush into position
  • Simple function
  • Tool
Specifications
Type LC Piston Seal Insertion Tool
For Use With (Application) Used to Simplifying Pump Maintenance by Seal Insertion and Cannot Mar The Surrounding Metal Surface of The Pump Housing
Quantity 1 Ea.