Learn More
Restek™ Injector Mounting Posts for Agilent Autosamplers
SDP
Catalog No. 06802654 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Packaging Quantity:
1 Each
Catalog No. Packaging Quantity
06-802-654 1 Each
1 options
Catalog No. 06-802-654 Supplier Restek™ Supplier No. 21237
For Agilent 7673 Series Autosamplers.
  • For use with 6890 GCs
Specifications
Type Injector Mounting Post
Packaging Quantity 1 Each
For Use With Agilent 7673 Series Autosamplers; 6890 GCs