Learn More
Restek™ Injector Wrench for Agilent 5890/6890/6850 GCs
SDP
Catalog No. 06711866 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Agilent 5890/6890/6850 GCs
Catalog No. For Use With (Equipment)
06-711-866 Agilent 5890/6890/6850 GCs
1 options
Catalog No. 06-711-866 Supplier Restek™ Supplier No. 22065
For Agilent 5890, 6850 and 6890 GCs.
  • Use to remove the septum nut and weldments during GC maintenance
  • Use the smaller end to remove the septum nut
  • Use the larger end to tighten the split/splitless weldment nut
  • High-quality stainless steel construction
Specifications
Type Inlet Wrench
Material Stainless Steel
For Use With (Equipment) Agilent 5890/6890/6850 GCs