Learn More
Restek™ Inlet Liner Packing Tool
SDP
Catalog No. 06710590 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
06-710-590 Each
1 options
Catalog No. 06-710-590 Supplier Restek™ Supplier No. 20339
Add to Cart
Edge
Add to Cart
Positions wool reproducibly every time.
  • Accurate to a specific, measured depth
  • Can be used with all manufacturers liners
  • Recommended for inlet liners with an ID ≥2mm
Specifications
Product Type Inlet Liner Packing Tool
Quantity Each
For Use With (Equipment) Liner