Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Inlet Liner Seals for Varian 1177 Injectors
SDP

Catalog No. p-3186657 Shop All Restek Corporation Products
Click to view available options
Quantity:
10/Pk.
50/Pk.
Diameter (Metric) Inner:
0.635 cm
0.65 cm

For Agilent and Bruker/Varian 1177 Injectors.

  • High thermal stability at injection port temperatures up to 450°C
  • For split or splitless liners

Specifications

Product Type O-ring
Quantity 50/Pk.
Temperature (Metric) Max 450°C
Diameter (Metric) Inner 0.635 cm
For Use With (Equipment) Split Liners, Agilent and Bruker/Varian 1177 Injectors
Liner Material Graphite
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.