Learn More
Restek™ Maintenance Supplies for Capillary Columns
SDP
Catalog No. 06710497 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With:
Capillary Columns
Catalog No. For Use With
06-710-497 Capillary Columns
1 options
Catalog No. 06-710-497 Supplier Restek™ Supplier No. 20101
Lock capillary columns to the correct insertion distance recommended by the instrument manufacturer during installation. Restek™ Slide-Lock Tweezers and 15cm Compact Steel Ruler are useful for many laboratory tasks.
  • An alternative way to install capillary columns to the correct insertion distance recommended by the instrument manufacturer
  • Useful for many laboratory tasks
Specifications
For Use With (Application) Capillary Column Installation
Description Slide-Lock Tweezers and 15 cm Compact Steel Ruler
For Use With Capillary Columns
Overall Length (Metric) 15 cm
Material Steel