Learn More
Restek™ MINICYL Regulator
SDP
Catalog No. 06710846 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Includes:
1/4 in. Tube Fittings
1/8 in. Tube Fittings
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
06-710-846 1/4 in. Tube Fittings
06-710-845 1/8 in. Tube Fittings
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-710-846 Supplier Restek™ Supplier No. 20611
Includes
0 to 60 psig (0 to 414kPa) gauge
Compact general purpose regulator
  • Laboratory use for air-drying glassware, sparging or evaporating solutions, and controlling pneumatic valves
  • Constructed of lightweight aluminum with an elastomer diaphragm
  • Choice of 0.32 or 0.64cm (0.125 or .25 in.) tube fittings
Specifications
Type Fitting
Includes 1/4 in. Tube Fittings
For Use With Restek MINICYL