Learn More
Restek™ MSD Conversion Fitting - Improved
SDP
Catalog No. 06711516 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
Fitting
Seal
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
06-711-516 Fitting
06-718-491 Seal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-711-516 Supplier Restek™ Supplier No. 21314
Add to Cart
Add to Cart
Flat, soft aluminum sealing ring deforms and butt-seals against the MSD interface.
  • Standard Vespel ferrule seals the column and 1/16 in. stainless steel nut
  • Constructed of nickel-plated brass for longevity and softness
  • Use any standard Vespel or Vespel/graphite 1/16 in. ferrule
  • Includes a 1/16 in. stainless steel nut and two replacement sealing rings
  • Improved design reduces chance of leaks
Specifications
Type Fitting
For Use With Chromatography Columns and Supplies