Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ MSD Source Nut
SDP

Catalog No. 06710868 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Material:
Brass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
06-710-868 Brass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-710-868 Supplier Restek™ Supplier No. 20643
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nut with 1.2mm bore permits easy removal of ferrules with a standard tapered-needle file.

  • Made of brass to prevent thread-stripping on the transfer line
  • Design enhances ease of threading onto the transfer line and improves overall lifetime
  • Quantity: 2/Pk.

Specifications

Diameter (Metric) Inner 1.2 mm
Material Brass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.