Learn More
Restek™ MXT-Union Connector Kits For Fused Silica Columns
SDP
Catalog No. 06711568 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With:
0.25 mm I.D. Fused Silica Columns
0.32 mm I.D. Fused Silica Columns
0.53 mm I.D. Fused Silica Columns
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
06-711-568 0.25 mm I.D. Fused Silica Columns
06-711-566 0.53 mm I.D. Fused Silica Columns
06-711-567 0.32 mm I.D. Fused Silica Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-711-568 Supplier Restek™ Supplier No. 21386
For connecting metal and/or fused silica GC columns.
  • Low dead volume, leak-tight connection
  • Reusable
  • Siltek treatment ensures maximum inertness
  • Ideal for connecting a guard column or transfer line to an analytical column
  • Use to oven temperatures of 350°C
  • Can also be used for connecting fused silica to metal
  • Includes MXT union, two 1/32 in. nuts and two one-piece, fused silica adapters
Specifications
Type Connector
Includes MXT union, two 1/32 in. nuts and two one-piece, fused silica adapters
For Use With 0.25 mm I.D. Fused Silica Columns