Learn More
Restek™ Open-End Wrench Sets
SDP
Catalog No. p-3186833 Shop All Restek Corporation Products
Change view
Click to view available options
For Use With:
Shimadzu 17 A, 2010, 2014 Capillary Installation Gauge
Tightening Chromatography Fittings
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
06-710-604 Tightening Chromatography Fittings
06-729-308 Shimadzu 17 A, 2010, 2014 Capillary Installation Gauge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-710-604 Supplier Restek™ Supplier No. 20387
Only null left
Add to Cart
Add to Cart
For tightening a wide variety of chromatography fittings
Model 20387 Set:
  • 1/4 in. x 5/16 in.
  • 3/8 in. x 7/16 in.
  • 7/16 in. x 1/2 in.
  • 1/2 in. x 9/16 in.
Model 22334 Set (For use with Shimadzu 17A, 2010 and 2014 Capillary Installation Gauge):
  • 1/4 in. x 5/16 in.
  • 10mm x 11mm (0.39 in. x 0.43 in.)
Specifications
Type Open End Wrench Set
For Use With Tightening Chromatography Fittings