Learn More
Restek™ PEEK™ Fitting Extractor
SDP
Catalog No. 06712865 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Material:
PEEK
Catalog No. Material
06-712-865 PEEK
1 options
Catalog No. 06-712-865 Supplier Restek™ Supplier No. 25325
Simple to use fitting extractor easily removes fitting without damaging the column.
  • Made from PEEK
  • Simply drill into the broken fitting, then screw the extractor into the fitting and remove it
  • Eliminates need for heat or other techniques
Specifications
Type Fitting
For Use With (Application) To Remove Broken Fitting
Material PEEK