Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ PEEK™ Tubing Elbows
SDP

Catalog No. 06712852 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
Angle Tubing Elbow, 180°
Angle Tubing Elbow, 90°
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
06-712-852 Angle Tubing Elbow, 180°
06-712-851 Angle Tubing Elbow, 90°
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-712-852 Supplier Restek™ Supplier No. 25309
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal for routing 1/16 in. PEEK tubing through your system.

  • Simply snap the tubing into the elbow
  • Prevent pinching of PEEK tubing, which can cause high pressure
  • Quantity: 5/Pk.

Specifications

Type Angle Tubing Elbow, 180°
Diameter (English) Inner 0.0625 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.