Learn More
Restek™ Pocket Magnifier
SDP
Catalog No. 06710504 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Application):
10X Magnification
Catalog No. For Use With (Application)
06-710-504 10X Magnification
1 options
Catalog No. 06-710-504 Supplier Restek™ Supplier No. 20124
Small and easy to handle
10x magnification makes it easy to see the column end to verify a square cut.
Specifications
Type Pocket Magnifier
For Use With (Application) 10X Magnification