Learn More
Restek™ Porous Polymers: HayeSep Series
SDP
Catalog No. 06712959
Click to view available options
Particle Size:
100/120 Mesh
60/80 Mesh
80/100 Mesh
Material:
Hayesep A
Hayesep B
Hayesep C
Hayesep D
Hayesep DB
Hayesep DIP
Hayesep N
Hayesep Q
Hayesep S
Hayesep T
Porous polymer adsorbent packing for GC columns.
Produce lower bleed and more reproducible characteristics.
Specifications
Particle Size 60/80 Mesh
Quantity 24 g
Material Hayesep D
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.