Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Porous Polymers: HayeSep Series
SDP

Catalog No. 06712962 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Particle Size:
100/120 Mesh
60/80 Mesh
80/100 Mesh
Material:
Hayesep A
Hayesep B
Hayesep C
Hayesep D
Hayesep DB
Hayesep DIP
Hayesep N
Hayesep Q
Hayesep S
Hayesep T
25 product options available for selection
Product selection table with 25 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Particle Size Material
06-712-962 100/120 Mesh Hayesep DIP
06-712-607 80/100 Mesh Hayesep A
06-712-608 100/120 Mesh Hayesep A
06-712-614 100/120 Mesh Hayesep D
06-712-613 80/100 Mesh Hayesep D
06-712-619 80/100 Mesh Hayesep N
06-712-620 100/120 Mesh Hayesep N
06-712-624 100/120 Mesh Hayesep Q
06-712-628 100/120 Mesh Hayesep S
06-712-630 100/120 Mesh Hayesep T
06-712-629 80/100 Mesh Hayesep T
06-712-959 60/80 Mesh Hayesep D
06-712-964 80/100 Mesh Hayesep DB
06-712-965 100/120 Mesh Hayesep DB
06-712-966 60/80 Mesh Hayesep N
06-712-968 60/80 Mesh Hayesep Q
06-712-971 60/80 Mesh Hayesep T
06-712-610 100/120 Mesh Hayesep B
06-712-609 80/100 Mesh Hayesep B
06-712-623 80/100 Mesh Hayesep Q
06-712-957 60/80 Mesh Hayesep B
06-712-958 60/80 Mesh Hayesep C
06-712-961 80/100 Mesh Hayesep DIP
06-712-960 60/80 Mesh Hayesep DIP
06-712-963 60/80 Mesh Hayesep DB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
25 options
Catalog No. 06-712-962 Supplier Restek™ Supplier No. 25566

Porous polymer adsorbent packing for GC columns.

Produce lower bleed and more reproducible characteristics.

Specifications

Particle Size 100/120 Mesh
Quantity 24 g
Material Hayesep DIP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.