Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Pre-Cleaned VOA Sampling Vials
SDP

Catalog No. p-3186876 Shop All Restek Corporation Products
Change view
Click to view available options
Volume (Metric):
20 mL
40 mL
Color:
Amber
Clear
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
06-718-679 40 mL Clear
06-718-681 20 mL Clear
06-718-680 40 mL Amber
06-718-682 20 mL Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-718-679 Supplier Restek™ Supplier No. 21796
Only null left
Add to Cart
Add to Cart

A container, liner and closure which are cleaned, assembled and lot traceable

  • Open top caps
  • PTFE faced 0.125 in. silicone septa

Compliance

Cleaned to U.S. EPA Protocol B specifications

Specifications

Color Clear
Material Glass
Volume (Metric) 40 mL
Closure Type Screw Cap (Open-Top)
Quantity 72 Pk.
Closure Size 24-400
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.