Learn More
Restek™ Protocol Station
SDP
Catalog No. 06711539 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Connection Type:
CGA 350 (H2, P5)
CGA 580 (N2, He, Ar)
CGA 590 (air)
Material:
Chrome-plated Brass
Stainless Steel
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type Material
06-711-539 CGA 580 (N2, He, Ar) Chrome-plated Brass
06-711-522 CGA 580 (N2, He, Ar) Stainless Steel
06-711-541 CGA 590 (air) Chrome-plated Brass
06-711-540 CGA 350 (H2, P5) Chrome-plated Brass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-711-539 Supplier Restek™ Supplier No. 21347
Add to Cart
Edge
Add to Cart
Requires
Pressure regulator
Designed for convenient wall mounting of high-purity gas regulators
Wall mounting of the protocol station provides ease of use, prevents gas regulator damage and improves safety.
  • With a 3 foot, flexible, all-stainless-steel pigtail with armor casing
  • CGA connection with an integral check valve
Specifications
Connection Type CGA 580 (N2, He, Ar)
Type Protocol Station
For Use With (Application) Wall mounting of the protocol station provides ease of use, prevents gas regulator damage and improves safety
For Use With (Equipment) Single or dual stage regulator
Includes 3 ft., Flexible, all Stainless Steel pigtail with armor casing
Material Chrome-plated Brass