Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Replacement CarboFrit Inserts
SDP

Catalog No. 06717674 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With:
Sleeves ≤4 mm I.D.
Sleeves Greater Than 4 mm I.D.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
06-717-674 Sleeves ≤4 mm I.D.
06-717-673 Sleeves Greater Than 4 mm I.D.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-717-674 Supplier Restek™ Supplier No. 20295
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CarboFrit plugs with OD slightly larger than 4mm ID of glass liners conform to the ID of the liner thereby preventing voids in the packing.

Part No. 20294

  • For Liner ID >4mm
  • Quantity: 10/Pk.

Part No. 20295

  • For Liner ID ≤4mm
  • Quantity: 10/Pk.

Specifications

Type Frit
Packaging Quantity 10 Pack
For Use With Sleeves ≤4 mm I.D.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.