Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ Replacement Frits for Passive Air Sampling Kits

For use in critical orifice fitting.
$92.00
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
06-712-234
![]() |
Restek™
24171 |
Siltek Treated |
Each of 1 for $92.00
|
|
|||||
Description
- Size: 2μm
- Available in stainless-steel or Siltek-treated material
- Includes washers
Specifications
Replacement Frit Filter | |
Washers | |
2 μm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title