Restek™ Replacement Frits for Passive Air Sampling Kits

For use in critical orifice fitting.

Manufacturer: Restek™ 24171

 View more versions of this product

Catalog No. 06-712-234

  Check Availability
Add to Cart

    Specifications

    Includes Washers
    Material Siltek™ treated
    Size 2μm
    Product Type Replacement Frit Filter

    • Size: 2µm
    • Available in stainless-steel or Siltek-treated material
    • Includes washers

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit