Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Replacement Inlet Cross-Disk Seals for Agilent 6890 GCs
SDP

Catalog No. 06717778 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Material:
Gold-plated
Siltek
Packaging Quantity:
10 Pack
2 Pack
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Packaging Quantity
06-717-778 Siltek 2 Pack
06-717-777 Siltek 10 Pack
06-717-779 Gold-plated 10 Pack
06-717-780 Gold-plated 2 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-717-778 Supplier Restek™ Supplier No. 20475
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Ideal for high-flow split applications >500mL/min. on Agilent 5890 GCs.

  • Siltek treatment provides enhanced inertness versus stainless steel
  • All seals include washers
  • I.D.: 0.8mm

Specifications

Type Seal
Diameter (Metric) Inner 0.8 mm
Packaging Quantity 2 Pack
For Use With Agilent 6890 GCs
Material Siltek
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.