Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Replacement Inlet Seals for Agilent 5890/6890 Split/Splitless Injection Ports
SDP

Catalog No. 06718489 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Material:
Gold-plated
Siltek
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
06-718-489 Siltek
06-718-487 Gold-plated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-718-489 Supplier Restek™ Supplier No. 21307
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Agilent GCs.

  • Siltek treatment provides enhanced inertness versus stainless steel
  • Include washers
  • Available Siltek or gold-plated seals
  • I.D.: 1.2mm

Note
The 1.2mm inlet seal is recommended when installing two columns using a two-hole Vespel/graphite ferrule.

Specifications

Type Seal
Diameter (Metric) Inner 1.2 mm
Packaging Quantity 2 Pack
For Use With Agilent 5890/6890 Split/Splitless Injection Ports
Material Siltek
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.