Learn More
Restek™ SGE™ Replacement Needles for Agilent Autosampler Syringes
SDP
Catalog No. 06712494 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Needle Gauge:
26 G
Catalog No. Needle Gauge
06-712-494 26 G
1 options
Catalog No. 06-712-494 Supplier Restek™ Supplier No. 24820
Use these replacement needles with Agilent 7673, 7683, 7693A, and 6850 Autosampler Syringes.
  • Do not reuse PTFE washers
Specifications
Description SGE
Needle Gauge 26 G
Product Type Replacement Needle
Volume (Metric) 0.5 μL
Needle Point Style Cone