Description
- Innovative bonding chemistry for batch-to-batch reproducibility, excellent thermal stability, and long life
- Wide range of bonded phases available
- Equivalent to Durapak and Porasil packings
Order Info
Note: There is a Minimum Order Quantity of 10.
Specifications
Specifications
| Particle Size | 80/100 Mesh |
| Type | Packing Material |
| Material | ResSil C |