Learn More
Restek™ Rethreading Tools
SDP
Catalog No. 06712125 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With:
1/16 in. Compression Fittings
1/4 in. Compression Fittings
1/8 in. Compression Fittings
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
06-712-125 1/4 in. Compression Fittings
06-712-123 1/16 in. Compression Fittings
06-712-124 1/8 in. Compression Fittings
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-712-125 Supplier Restek™ Supplier No. 23018
Repair worn or damaged threads
  • Multiple uses (fittings, injectors, detectors, etc.)
  • Built-in guide to prevent cross-threading
Specifications
Type Rethreading Tool
For Use With 1/4 in. Compression Fittings