Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Rough Pump Oil
SDP

Catalog No. 06712029 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Packaging Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
06-712-029 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-712-029 Supplier Restek™ Supplier No. 22687
May include imposed supplier surcharges.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

For MSD and Oil Vacuum Pumps.

  • Formulated from crude oil stocks known for their durability and line-lubricating qualities
  • Use in Agilent 5973, 5972, 5971 and GCD mass spec systems
  • Under average use, the oil in the foreline rough pump should be replaced every six months
  • Quantity: 1L

Specifications

Type Oil Pump
Packaging Quantity 1 L
For Use With Agilent 5973, 5972, 5971 and GCD Mass Spec Systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.