Learn More
Description
Using sophisticated computer modeling software, Restek created two stationary phases for separating the 53 organophosphorus pesticides (OPP) listed in EPA Method 8141. The extended upper temperature limit of these phases (330°C) allows analysts to bake out high molecular weight contamination typically associated with pesticide samples. The low-bleed columns are a perfect match for sensitive detection systems.
- Proprietary Crossbond phases
- Fused silica
- Low bleed makes it ideal for GC-FPD, GC-NPD, or GC-MS analyses
- Improved separation and reduced analysis time
- Stable to 330° to 340°C
Order Info
Custom lengths and film thicknesses available.
Specifications
Specifications
| For Use With (Application) | Organochlorine Pesticides Analysis, GC/NPD or GC/MS, Organochlorine Pesticides, Herbicides Analysis |
| Film Thickness | 0.25 μm |
| Length (Metric) | 30 m |
| Material | Stainless Steel |
| Coating | Fused Silica |
| USP Type | None |
| Stationary Phase | Rtx™-OPP |
| Type | Capillary Column |
| Diameter (Metric) Inner | 0.25 mm |
| Max. Temperature (Metric) | 350°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
